1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate
OR308069-1G OR308069-5G OR308069-25G
APOSOR308069-1GEA 52.2 EUR
APOSOR308069-1G APOSOR308069-5G APOSOR308069-25G
1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate 97%
1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate
Formula: C₁₁H₂₀N₂O₄
MW: 244.29 g/mol
Melting Pt: 159…160 °C
Storage Temperature: Ambient
MDL Number: MFCD01632465
CAS Number: 129799-08-2