2,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Diazaspiro[3.4]octane, N6-BOC protected, tert-Butyl 2,6-diazaspiro[3.4]octane-6-carboxylate
OR308111-100MG OR308111-1G OR308111-250MG
APOSOR308111-100MGEA 58 EUR
APOSOR308111-100MG APOSOR308111-1G APOSOR308111-250MG
2,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester
2,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester
Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08234738
CAS Number: 885270-86-0