4-Cyanobenzamide 98%
Supplier: Apollo Scientific
Warning
OR310641-5G OR310641-25G
APOSOR310641-5GEA 95.7 EUR
APOSOR310641-5G APOSOR310641-25G
4-Cyanobenzamide 98%
4-Cyanobenzamide
Formula: C₈H₆N₂O
Melting Pt: 225…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00017133
CAS Number: 3034-34-2