2-Iodoxybenzoic acid
Supplier: Apollo Scientific
Danger
Synonyms: 1-Hydroxy-1,2-benziodoxy-3-(1h)-one-1-xoide, IBX, 2-Iodylbenzoic acid
OR3108-100G OR3108-1G OR3108-25G
APOSOR3108-100GEA 201 EUR
APOSOR3108-100G APOSOR3108-1G APOSOR3108-25G
2-Iodoxybenzoic acid
2-Iodoxybenzoic acid
Formula: C₇H₅IO₄
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02912492
CAS Number: 61717-82-6
UN: 1759
ADR: 8,II