N-(5-Bromo-2-pyridyl)thiourea
Supplier: Apollo Scientific
OR311004-1G OR311004-5G
APOSOR311004-1GEA 127 EUR
APOSOR311004-1G APOSOR311004-5G
N-(5-Bromo-2-pyridyl)thiourea
N-(5-Bromo-2-pyridyl)thiourea
Formula: C₆H₆BrN₃S
MW: 232.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15525454
CAS Number: 31430-38-3