4-Bromocrotonic acid 95%, Technical Grade
Supplier: Apollo Scientific
Danger
Synonyms: trans-4-Bromo-2-butenoic Acid, (E)-4-Bromobut-2-enoic acid
OR313099-5G OR313099-25G
APOSOR313099-5GEA 65.4 EUR
APOSOR313099-5G APOSOR313099-25G
4-Bromocrotonic acid 95%, Technical Grade
4-Bromocrotonic acid
Formula: C₄H₅BrO₂
Melting Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082701
CAS Number: 13991-36-1
UN: 3261
ADR: 8,III