(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Supplier: Apollo Scientific
Warning
Synonyms: (R)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide, (R)-(-)-1,1'-Binaphthalene-2,2'-diyl-phosphate
OR315004-25G OR315004-5G
APOSOR315004-25GEA 115 EUR
APOSOR315004-25G APOSOR315004-5G
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Formula: C₂₀H₁₃O₄P
MW: 348.289 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010045
CAS Number: 39648-67-4