4-Chloropyridine-N-oxide 95%
Supplier: Apollo Scientific
Warning
OR315222-1G OR315222-5G OR315222-25G
APOSOR315222-1GEA 79.9 EUR
APOSOR315222-1G APOSOR315222-5G APOSOR315222-25G
4-Chloropyridine-N-oxide 95%
4-Chloropyridine N-oxide
Formula: C₅H₄ClNO
Melting Pt: 160 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047425
CAS Number: 1121-76-2
EINECS: 214-336-2