Tetrahydro-3-furanylmethyl bromide 95%
Supplier: Apollo Scientific
Synonyms: 3-(Bromomethyl)tetrahydrofuran
OR315500-1G OR315500-5G
APOSOR315500-1GEA 178 EUR
APOSOR315500-1G APOSOR315500-5G
Tetrahydro-3-furanylmethyl bromide 95%
Tetrahydro-3-furanylmethyl bromide
Formula: C₅H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD08234790
CAS Number: 165253-29-2