8-Hydroxyoctanoic acid
Supplier: Apollo Scientific
Danger
OR315581-1G OR315581-250MG
APOSOR315581-1GEA 0 EUR
APOSOR315581-1G APOSOR315581-250MG
8-Hydroxyoctanoic acid
8-Hydroxyoctanoic acid
Formula: C₈H₁₆O₃
Melting Pt: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00792446
CAS Number: 764-89-6
UN: 3261
ADR: 8,II