1,5-Bis(diphenylphosphino)pentane 97%
Supplier: Apollo Scientific
Synonyms: Pentane-1,3-diylbis(diphenylphosphine), Pentylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)pentane
OR315706-25G OR315706-10G
APOSOR315706-25GEA 0 EUR
APOSOR315706-25G APOSOR315706-10G
1,5-Bis(diphenylphosphino)pentane 97%
1,5-Bis(diphenylphosphino)pentane
Formula: C₂₉H₃₀P₂
MW: 440.5 g/mol
Boiling Pt: 230
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00003052
CAS Number: 27721-02-4