Bis[2-(diphenylphosphino)phenyl]ether 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2'-Bis(diphenylphosphino)diphenyl Ether, DPEphos, Bis(2-diphenylphosphinophenyl)ether, (Oxydi-2,1-phenylene)bis(diphenylphosphine), (Oxybis(2,1-phenylene))bis(diphenylphosphine)
OR315707-25G OR315707-100G OR315707-5G
APOSOR315707-25GEA 36.4 EUR
APOSOR315707-25G APOSOR315707-100G APOSOR315707-5G
Bis[2-(diphenylphosphino)phenyl]ether 95+%
Bis[2-(diphenylphosphino)phenyl]ether
Formula: C₃₆H₂₈OP₂
MW: 538.57 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00233863
CAS Number: 166330-10-5