1-Cyclobutyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Supplier: Apollo Scientific
Warning
Synonyms: 1-Cyclobutyl-1H-pyrazole-4-boronic acid pinacol ester
OR317114-1G OR317114-5G
APOSOR317114-1GEA 59.6 EUR
APOSOR317114-1G APOSOR317114-5G
1-Cyclobutyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
1-Cyclobutyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formula: C₁₃H₂₁BN₂O₂
MW: 248.1290 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16659010
CAS Number: 1002309-48-9