2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one 95+%
Supplier: Apollo Scientific
Synonyms: 7-Deazaguanine
OR318028-1G OR318028-25G OR318028-5G
APOSOR318028-1GEA 52.2 EUR
APOSOR318028-1G APOSOR318028-25G APOSOR318028-5G
2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one 95+%
2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
Formula: C₆H₆N₄O
MW: 150.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09033259
CAS Number: 7355-55-7