Methyl-4-((4-oxopiperidin-1-yl)methyl)benzoate 95+%
Supplier: Apollo Scientific
OR318061-1G OR318061-10G OR318061-5G
APOSOR318061-1GEA 229 EUR
APOSOR318061-1G APOSOR318061-10G APOSOR318061-5G
Methyl-4-((4-oxopiperidin-1-yl)methyl)benzoate 95+%
Methyl-4-((4-oxopiperidin-1-yl)methyl)benzoate
Formula: C₁₄H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD11052315
CAS Number: 943767-91-7