8-Bromo-5-chloro-1,6-naphthyridine 95+%
Supplier: Apollo Scientific
Warning
OR318081-1G OR318081-5G OR318081-10G
APOSOR318081-1GEA 1210 EUR
APOSOR318081-1G APOSOR318081-5G APOSOR318081-10G
8-Bromo-5-chloro-1,6-naphthyridine 95+%
8-Bromo-5-chloro-1,6-naphthyridine
Formula: C₈H₄BrClN₂
Melting Pt: 128…129 °C
Storage Temperature: Ambient
MDL Number: MFCD08567949
CAS Number: 909649-12-3