2-(Oxiran-2-ylmethoxy)benzonitrile 95+%
Supplier: Apollo Scientific
OR318126-1G OR318126-10G OR318126-5G
APOSOR318126-1GEA 318 EUR
APOSOR318126-1G APOSOR318126-10G APOSOR318126-5G
2-(Oxiran-2-ylmethoxy)benzonitrile 95+%
2-(Oxiran-2-ylmethoxy)benzonitrile
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01632505
CAS Number: 38465-16-6