tert-Butyl (pyrrolidin-2-ylmethyl)carbamate 95%
Supplier: Apollo Scientific
Synonyms: 2-Boc-aminomethyl-pyrrolidine
OR321459-5G OR321459-25G
APOSOR321459-5GEA 133 EUR
APOSOR321459-5G APOSOR321459-25G
tert-Butyl (pyrrolidin-2-ylmethyl)carbamate 95%
tert-Butyl (pyrrolidin-2-ylmethyl)carbamate
Formula: C₁₀H₂₀N₂O₂
MW: 200.2780 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658352
CAS Number: 149649-58-1