7-Hydroxyquinolin-2(1H)-one 97%

Supplier: Apollo Scientific

OR321481-25G OR321481-5G
APOSOR321481-25GEA 45.9 EUR
APOSOR321481-25G APOSOR321481-5G
7-Hydroxyquinolin-2(1H)-one 97%
7-Hydroxyquinolin-2(1H)-one
Formula: C₉H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD07644575
CAS Number: 70500-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR