Iodoacetic acid
Supplier: Apollo Scientific
Danger
OR322006-100G OR322006-25G
APOSOR322006-100GEA 348 EUR
APOSOR322006-100G APOSOR322006-25G
Iodoacetic acid
Iodoacetic acid
Formula: C₂H₃IO₂
Boiling Pt: 208 °C
Melting Pt: 79…82 °C
Density: 4.6 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00002685
CAS Number: 64-69-7
UN: 2923
ADR: 8+6.1,I