2,4-Dimethylthiazole
Supplier: Apollo Scientific
Warning
OR322034-100G OR322034-500G
APOSOR322034-100GEA 37.7 EUR
APOSOR322034-100G APOSOR322034-500G
2,4-Dimethylthiazole
2,4-Dimethylthiazole
Formula: C₅H₇NS
Boiling Pt: 144…146 °C
Density: 1.05 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00014509
CAS Number: 541-58-2
UN: 1993
ADR: 3,III