1-Naphthyl isothiocyanate 98%
Supplier: Apollo Scientific
OR322081-25G OR322081-5G
APOSOR322081-25GEA 340 EUR
APOSOR322081-25G APOSOR322081-5G
1-Naphthyl isothiocyanate 98%
1-Naphthyl isothiocyanate
Formula: C₁₁H₇NS
MW: 185.25 g/mol
Boiling Pt: 100
Melting Pt: 59
Density: 1.81
Storage Temperature: Ambient
MDL Number: MFCD00003882
CAS Number: 551-06-4
Merck Index: 12,06498