(S)-(-)-2-Acetoxypropionic acid
Supplier: Apollo Scientific
Warning
OR322114-1G OR322114-5G
APOSOR322114-1GEA 21.5 EUR
APOSOR322114-1G APOSOR322114-5G
(S)-(-)-2-Acetoxypropionic acid
(S)-(-)-2-Acetoxypropionic acid
Formula: C₅H₈O₄
Boiling Pt: 142 °C/20mmHg
Density: 1.18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00191370
CAS Number: 6034-46-4