N,N-Diisopropylcarbamoyl chloride
Supplier: Apollo Scientific
Danger
OR322162-25G OR322162-5G
APOSOR322162-25GEA 121 EUR
APOSOR322162-25G APOSOR322162-5G
N,N-Diisopropylcarbamoyl chloride
N,N-Diisopropylcarbamoyl chloride
Formula: C₇H₁₄ClNO
Boiling Pt: 90…93 °C/15mmHg
Melting Pt: 55…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00015013
CAS Number: 19009-39-3
EINECS: 242-743-5
UN: 3261
ADR: 8,II