4-Nitropyrocatechol

Supplier: Apollo Scientific

Synonyms: 4-Nitrocatechol, 4-Nitrobenzene-1,2-diol

OR322229-100G OR322229-25G
APOSOR322229-100GEA 469 EUR
APOSOR322229-100G APOSOR322229-25G
4-Nitropyrocatechol
4-Nitropyrocatechol
Formula: C₆H₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00007242
CAS Number: 3316-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR