(S)-(-)-4-Bromo-α-methylbenzylamine
Supplier: Apollo Scientific
Danger
OR322311-5G OR322311-25G
APOSOR322311-5GEA 0 EUR
APOSOR322311-5G APOSOR322311-25G
(S)-(-)-4-Bromo-α-methylbenzylamine
(S)-(-)-4-Bromo-α-methylbenzylamine
Formula: C₈H₁₀BrN
Boiling Pt: 140…145 °C/30mmHg
Melting Pt: –25 °C
Density: 1.39 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066026
CAS Number: 27298-97-1
UN: 3265
ADR: 8,III