1,4-Diazabicyclo[2.2.2]octane
Supplier: Apollo Scientific
Danger
Synonyms: DABCO
OR322352-1KG OR322352-500G OR322352-100G
APOSOR322352-1KGEA 69.6 EUR
APOSOR322352-1KG APOSOR322352-500G APOSOR322352-100G
1,4-Diazabicyclo[2.2.2]octane
1,4-Diazabicyclo[2.2.2]octane
Formula: C₆H₁₂N₂
MW: 112.17 g/mol
Boiling Pt: 174 °C/760mmHg
Melting Pt: 156…159 °C
Density: 1.02 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00006689
CAS Number: 280-57-9
UN: 1325
ADR: 4.1 II
Merck Index: 12,09801