3-Acetylaminobenzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
OR322436-1G OR322436-250MG
APOSOR322436-1GEA 339 EUR
APOSOR322436-1G APOSOR322436-250MG
3-Acetylaminobenzenesulphonyl chloride
3-Acetylaminobenzenesulphonyl chloride
Formula: C₈H₈ClNO₃S
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD02222873
CAS Number: 23905-46-6
UN: 3261
ADR: 8,III