5-tert-Butyl 3-ethyl 1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-3,5-dicarboxylate 95%
Supplier: Apollo Scientific
Warning
Synonyms: 5-tert-Butyl 3-ethyl 6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-3,5(4H)-dicarboxylate
OR322647-1G OR322647-250MG OR322647-5G
APOSOR322647-1GEA 120 EUR
APOSOR322647-1G APOSOR322647-250MG APOSOR322647-5G
5-tert-Butyl 3-ethyl 1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-3,5-dicarboxylate 95%
5-tert-Butyl-3-ethyl 1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-3,5-dicarboxylate
Formula: C₁₄H₂₁N₃O₄
MW: 295.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12028385
CAS Number: 518990-23-3