6-Nitropyrrolo[1,2-a]quinoline 95%

Supplier: Apollo Scientific

OR32494-1G OR32494-5G
APOSOR32494-1GEA 336 EUR
APOSOR32494-1G APOSOR32494-5G
6-Nitropyrrolo[1,2-a]quinoline 95%
6-Nitropyrrolo[1,2-a]quinoline
Formula: C₁₂H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03791182
CAS Number: 52414-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR