4-Bromo-2-furaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromofurfural, 4-Bromofuran-2-carbaldehyde
OR3267-1G OR3267-250MG OR3267-5G
APOSOR3267-1GEA 94.4 EUR
APOSOR3267-1G APOSOR3267-250MG APOSOR3267-5G
4-Bromo-2-furaldehyde
4-Bromo-2-furaldehyde
Formula: C₅H₃BrO₂
MW: 174.98 g/mol
Melting Pt: 54…58 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671504
CAS Number: 21921-76-6