1-Phenyl-1H-1,2,3-triazole-4-carbaldehyde 95%
Supplier: Apollo Scientific
OR32841-1G
OR32841-5G
APOSOR32841-1GEA
538
EUR
APOSOR32841-1G
APOSOR32841-5G
1-Phenyl-1H-1,2,3-triazole-4-carbaldehyde 95%
1-Phenyl-1H-1,2,3-triazole-4-carbaldehyde
Formula:
C₉H₇N₃O Melting Pt: 98.5…99…5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100219 CAS Number: 34296-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...