N-(2-Hydroxy-1,1-dimethylethyl)-2-methoxybenzenecarboxamide 95%
Supplier: Apollo Scientific
Synonyms: N-(1-Hydroxy-2-methylpropan-2-yl)-2-methoxybenzamide
OR32984-1G OR32984-5G
APOSOR32984-1GEA 270 EUR
APOSOR32984-1G APOSOR32984-5G
N-(2-Hydroxy-1,1-dimethylethyl)-2-methoxybenzenecarboxamide 95%
N-(2-Hydroxy-1,1-dimethylethyl)-2-methoxybenzenecarboxamide
Formula: C₁₂H₁₇NO₃
MW: 223.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00665208
CAS Number: 74201-13-1