trans-4-Aminocyclohexanecarboxylic acid hydrochloride
Supplier: Apollo Scientific
Warning
OR3299-1G OR3299-5G
APOSOR3299-1GEA 21.5 EUR
APOSOR3299-1G APOSOR3299-5G
trans-4-Aminocyclohexanecarboxylic acid hydrochloride
trans-4-Aminocyclohexanecarboxylic acid hydrochloride
Formula: C₇H₁₄ClNO₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD03844587
CAS Number: 27960-59-4