Methyl-1-cyanocyclopentanecarboxylate 95%
Supplier: Apollo Scientific
OR33519-1G OR33519-5G
APOSOR33519-1GEA 270 EUR
APOSOR33519-1G APOSOR33519-5G
Methyl-1-cyanocyclopentanecarboxylate 95%
Methyl-1-cyanocyclopentanecarboxylate
Formula: C₈H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD09972231
CAS Number: 40862-12-2