3-Benzyloxy-4-methylphenylamine

Supplier: Apollo Scientific
Warning

OR346095-1G OR346095-5G
APOSOR346095-1GEA 121 EUR
APOSOR346095-1G APOSOR346095-5G
3-Benzyloxy-4-methylphenylamine
3-Benzyloxy-4-methylphenylamine
Formula: C₁₄H₁₅NO
Boiling Pt: 157 °C/ 1mmHg
Storage Temperature: Ambient
MDL Number: MFCD04973042
CAS Number: 219492-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR