3-Ethynyltoluene
Supplier: Apollo Scientific
Warning
OR3481-1G OR3481-25G OR3481-5G
APOSOR3481-1GEA 21.9 EUR
APOSOR3481-1G APOSOR3481-25G APOSOR3481-5G
3-Ethynyltoluene
3-Ethynyltoluene
Formula: C₉H₈
Boiling Pt: 170…175 °C
Density: 0.9 g/cm³ (25 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD01318174
CAS Number: 766-82-5
UN: 3295
ADR: 3,III