4-Bromo-3-methyl-1H-pyrazole
Supplier: Apollo Scientific
Warning
OR350075-25G OR350075-5G
APOSOR350075-25GEA 0 EUR
APOSOR350075-25G APOSOR350075-5G
4-Bromo-3-methyl-1H-pyrazole
4-Bromo-3-methyl-1H-pyrazole
Formula: C₄H₅BrN₂
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00005241
CAS Number: 13808-64-5