5-Chloro-m-xylene 96%
Supplier: Apollo Scientific
Warning
OR350094-5G OR350094-1G OR350094-25G
APOSOR350094-5GEA 26.1 EUR
APOSOR350094-5G APOSOR350094-1G APOSOR350094-25G
5-Chloro-m-xylene 96%
5-Chloro-m-xylene
Formula: C₈H₉Cl
Boiling Pt: 187 °C
Density: 1.04 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00060273
CAS Number: 556-97-8