Methyl-3,4-dihydroxybenzoate
Supplier: Apollo Scientific
Warning
OR350236-100G OR350236-25G
APOSOR350236-100GEA 65.4 EUR
APOSOR350236-100G APOSOR350236-25G
Methyl-3,4-dihydroxybenzoate
Methyl 3,4-dihydroxybenzoate
Formula: C₈H₈O₄
Melting Pt: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00016437
CAS Number: 2150-43-8