3-Nitro-o-xylene
Supplier: Apollo Scientific
Warning
Synonyms: 1,2-Dimethyl-3-nitrobenzene, 2,3-Dimethylnitrobenzene, 3-Nitro-o-xylene
OR350286-500G OR350286-1KG OR350286-250G
APOSOR350286-500GEA 132 EUR
APOSOR350286-500G APOSOR350286-1KG APOSOR350286-250G
3-Nitro-o-xylene
3-Nitro-o-xylene
Formula: C₈H₉NO₂
Boiling Pt: 244…246 °C
Melting Pt: 13…15 °C
Density: 1.2129 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00007162
CAS Number: 83-41-0
UN: 1665
ADR: 6.1,II