DL-7-Azatryptophan monohydrate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-3-(7H-pyrrolo[2,3-b]pyridin-3-yl)propanoic acid, H-DL-7-Aza-Trp-OH
OR3563-1G OR3563-250MG
APOSOR3563-1GEA 598 EUR
APOSOR3563-1G APOSOR3563-250MG
DL-7-Azatryptophan monohydrate 97%
DL-7-Azatryptophan monohydrate
Formula: C₁₀H₁₁N₃O₂.H₂O
MW: 223.24 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037930
CAS Number: 7303-50-6