6-Bromoquinolin-4-ol
Supplier: Apollo Scientific
Warning
Synonyms: 6-Bromo-4-quinolinol, 6-Bromo-4-hydroxyquinoline
OR3631-1G OR3631-5G
APOSOR3631-1GEA 0 EUR
APOSOR3631-1G APOSOR3631-5G
6-Bromoquinolin-4-ol
6-Bromoquinolin-4-ol
Formula: C₉H₆BrNO
MW: 224.0540 g/mol
Melting Pt: 281…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00272435
CAS Number: 145369-94-4