(6-Bromo-2-pyridinyl)methanol 95%
Supplier: Apollo Scientific
Warning
Synonyms: (6-Bromopyridin-2-yl)methanol, 2-Bromo-6-pyridinemethanol, 2-Bromo-6-(hydroxymethyl)pyridine
OR3668-25G OR3668-5G
APOSOR3668-25GEA 192 EUR
APOSOR3668-25G APOSOR3668-5G
(6-Bromo-2-pyridinyl)methanol 95%
(6-Bromo-2-pyridinyl)methanol
Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 246 °C
Melting Pt: 34…39 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04039884
CAS Number: 33674-96-3