2-Amino-6-iodopurine
Supplier: Apollo Scientific
Warning
OR3725-1G OR3725-250MG OR3725-5G
APOSOR3725-1GEA 34.4 EUR
APOSOR3725-1G APOSOR3725-250MG APOSOR3725-5G
2-Amino-6-iodopurine
2-Amino-6-iodopurine
Formula: C₅H₄IN₅
Melting Pt: 240 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD08706375
CAS Number: 19690-23-4