2-Amino-6-methoxypurine

Supplier: Apollo Scientific
Warning

OR3726-100G OR3726-25G OR3726-5G
APOSOR3726-100GEA 171 EUR
APOSOR3726-100G APOSOR3726-25G APOSOR3726-5G
2-Amino-6-methoxypurine
2-Amino-6-methoxypurine
Formula: C₆H₇N₅O
Storage Temperature: Ambient
MDL Number: MFCD00133009
CAS Number: 20535-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR