4-Chlorotoluene 98%
Supplier: Apollo Scientific
Synonyms: 1-Chloro-4-methylbenzene
OR3751-1KG OR3751-500G
APOSOR3751-1KGEA 219 EUR
APOSOR3751-1KG APOSOR3751-500G
4-Chlorotoluene 98%
4-Chlorotoluene
Formula: C₇H₇Cl
Boiling Pt: 161…163 °C
Melting Pt: 44446 °C
Density: 1.069 g/cm³ (25 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00000631
CAS Number: 106-43-4