2,4-Dichlorotoluene
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Dichloro-4-methylbenzene
OR3753-1KG OR3753-500G
APOSOR3753-1KGEA 328 EUR
APOSOR3753-1KG APOSOR3753-500G
2,4-Dichlorotoluene
2,4-Dichlorotoluene
Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 199…201 °C
Melting Pt: –14…–13 °C
Density: 1.246 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000583
CAS Number: 95-73-8
EINECS: 202-445-8
UN: 3082
ADR: 9+EHS III