4-Cyanobenzoyl chloride 98%

Supplier: Apollo Scientific
Danger

OR3784-25G OR3784-5G
APOSOR3784-25GEA 82.8 EUR
APOSOR3784-25G APOSOR3784-5G
4-Cyanobenzoyl chloride 98%
4-Cyanobenzoyl chloride
Formula: C₈H₄ClNO
MW: 165.58 g/mol
Boiling Pt: 110 °C/2mmHg
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00001822
CAS Number: 6068-72-0
EINECS: 228-005-5
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR