2,5-Dichloronicotinic acid
Supplier: Apollo Scientific
Warning
OR3876-1G OR3876-5G
APOSOR3876-1GEA 21.9 EUR
APOSOR3876-1G APOSOR3876-5G
2,5-Dichloronicotinic acid
2,5-Dichloronicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD01861979
CAS Number: 59782-85-3